Learn More
1-Indanol, TRC
Product Code. 30135936
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135936 10 g 10g
30137760 5 g 5g
30010074 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135936 Supplier Toronto Research Chemicals Supplier No. TRCI49985310G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6351-10-6
Molecular Formula C9 H10 O
Molecular Weight (g/mol) 134.175
Synonym 1H-Inden-1-ol, 2,3-dihydro-, 1-Indanol (6CI,7CI,8CI), 1-Indanol, (±)-, 2,3-Dihydro-1H-inden-1-ol, (RS)-1-Indanol, (±)-1-Indanol, (±)-2,3-Dihydro-1H-inden-1-ol, 1-Hydroxyhydrindene, 1-Hydroxyindan, NSC 31258
IUPAC Name 2,3-dihydro-1H-inden-1-ol
SMILES OC1CCc2ccccc12
Specifications
Quantity 10 g
InChI Formula InChI=1S/C9H10O/c10-9-6-5-7-3-1-2-4-8(7)9/h1-4,9-10H,5-6H2
Formula Weight 134.0732
Chemical Name or Material 1-Indanol