Learn More
1-Hexene, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Hexene |
| CAS | 592-41-6 |
| Molecular Formula | C6 H12 |
| Quantity | 5 g |
| Synonym | 1-Hexene, 1-n-Hexene, Dialen 6, Hexene, Linealene 6, NSC 74121, α-Hexene |
| InChI Formula | InChI=1S/C6H12/c1-3-5-6-4-2/h3H,1,4-6H2,2H3 |
| SMILES | CCCCC=C |
| IUPAC Name | hex-1-ene |
| Molecular Weight (g/mol) | 84.16 |
| Formula Weight | 84.0939 |