Learn More
1-Heptanol, 99%
Product Code. 11485143
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
Product Code. Quantity unitSize
11485143 100 mL 100mL
11495143 500 mL 500mL
11405153 2500 mL 2500mL
3 options
Product Code. 11485143
Brand: Thermo Scientific Alfa Aesar A12793.AE
Request Bulk or Custom Format
Edge
CAS: 111-70-6 | C7H16O | 116.20 g/mol
1-Heptanol, is used in the fragrance industry due to its pleasant smell.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Heptanol, is used in the fragrance industry due to its pleasant smell.
Solubility
Soluble in water 1.7 g/L at 25°C.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 111-70-6
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00002986
InChI Key BBMCTIGTTCKYKF-UHFFFAOYSA-N
Synonym 1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
PubChem CID 8129
IUPAC Name heptan-1-ol
SMILES CCCCCCCO
Specifications
Melting Point -34°C
Density 0.82
Boiling Point 175°C to 176°C
Flash Point 73°C (163°F)
Assay Percent Range 99%
Odor Characteristic
Linear Formula CH3(CH2)6OH
Refractive Index 1.424
Quantity 100 mL
Beilstein 1731686
Merck Index 14,4661
Solubility Information Soluble in water 1.7g/L at 25°C.
Formula Weight 116.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Heptanol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Heptanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1-Heptanol