Learn More
1-Eicosanol, Sigma-Aldrich™
Product Code. 15841631
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15841631 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15841631 Supplier Sigma Aldrich Supplier No. 8220510100
 Restricted Item
Estimated Shipment: 20-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 629-96-9
Melting Point 65°C
Density 0.911 g/cm3 at 20°C
Molecular Formula CH3(CH2)19OH
MDL Number MFCD00002938
Quantity 100 g
Synonym 1-Eicosanol, Arachyl alcohol, 1-Eicosanol
Molecular Weight (g/mol) 298.55
Grade Research
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.