Learn More
1-Docosanol, 98%
Product Code. p-2689538 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
Product Code. Quantity Packaging unitSize
10272262 25 g Plastic bottle 25g
10581082 100 g Plastic bottle 100g
2 options
Product Code. 10272262
Brand: Thermo Scientific Acros 173720250
Request Bulk or Custom Format
CAS: 661-19-8 | C22H46O | 326.61 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 661-19-8
Molecular Formula C22H46O
Molecular Weight (g/mol) 326.61
MDL Number MFCD00002939
InChI Key NOPFSRXAKWQILS-UHFFFAOYSA-N
Synonym 1-docosanol, behenyl alcohol, docosanol, behenic alcohol, docosyl alcohol, n-docosanol, abreva, tadenan, lidavol, lanette 22
PubChem CID 12620
ChEBI CHEBI:31000
IUPAC Name docosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCO
Specifications
CAS Min % 97.5
CAS Max % 100
Melting Point 68.0°C to 72.0°C
Color White
Flash Point 227°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula CH3(CH2)21OH
Quantity 25 g
Beilstein 01, 431
Merck Index 15, 3444
Solubility Information Solubility in water: insoluble.
Viscosity 7.5 mPa.s (80°C)
Formula Weight 326.61
Percent Purity 98%
Physical Form Pellets or Tablets
Chemical Name or Material 1-Docosanol, 0.98