Learn More
1-Diazo-2-naphthol-4-sulfonic acid, 76%, Thermo Scientific™
Product Code. 15427308
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. 15427308
Brand: Thermo Scientific Alfa Aesar H60805.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 887-76-3
Molecular Formula C10H6N2O4S
MDL Number MFCD00012409
Specifications
Melting Point 160°C (decomposition)
Quantity 25 g
UN Number UN2925
Formula Weight 250.23
Percent Purity 76%
Chemical Name or Material 1-Diazo-2-naphthol-4-sulfonic acid