Learn More
1-Decanol, Sigma-Aldrich™
Product Code. 15879010
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15879010 1 L 1 L
15869010 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15879010 Supplier Sigma Aldrich Supplier No. 8034631000
 Restricted Item
Estimated Shipment: 24-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 112-30-1
Grade Research
Molecular Formula CH3(CH2)9OH
Molecular Weight (g/mol) 158.28
Specifications
Melting Point 5°C to 7°C
Density 0.8 g/cm3 at 20°C (DIN 51757)
Boiling Point 220°C to 235°C/1013 hPa
MDL Number MFCD00004747
Quantity 1 L
Synonym 1-Decanol, Capric alcohol, Decyl alcohol, 1-Decanol