Learn More
1-Cyano-4-(dimethylamino)pyridinium tetrafluoroborate, 98%
Product Code. 15424976
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. Quantity unitSize
15424976 500 mg 500mg
15414976 100 mg 100mg
2 options
Product Code. 15424976
Brand: Thermo Scientific Alfa Aesar H59334.ME
Request Bulk or Custom Format
CAS: 59016-56-7 | C8H10BF4N3 | 234.99 g/mol
1-Cyano-4-(dimethylamino)pyridinium tetrafluoroborate is a cyanylation reagent for protein sulfhydryl groups and used to prepare protein-polysaccharide conjugates. It is employed as an activating agent for polysaccharide resins and conjugation of lipopolysaccharides while retaining endotoxic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Cyano-4-(dimethylamino)pyridinium tetrafluoroborate is a cyanylation reagent for protein sulfhydryl groups and used to prepare protein-polysaccharide conjugates. It is employed as an activating agent for polysaccharide resins and conjugation of lipopolysaccharides while retaining endotoxic activity.
Solubility
Soluble in water, and acetonitrile.
Notes
Store at room temperature. Keep away from oxidizing agents and heats.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 59016-56-7
Molecular Formula C8H10BF4N3
Molecular Weight (g/mol) 234.99
MDL Number MFCD00011998
InChI Key MBLVMDCQDCVKNE-UHFFFAOYSA-N
Synonym cdap, 1-cyano-4-dimethylamino pyridinium tetrafluoroborate, 1-cyano-4-dimethylamino pyridin-1-ium tetrafluoroborate, unii-p4w72066jt, 1-cyano-4-dimethylamino-pyridinium tetrafluoroborate, pubchem17735, acmc-209m9a, n-cyano-4-dimethylaminopyridinium tetrafluoroborate, 1-cyano-4-dimethylaminopyridinium tetra-fluoroborate, n-cyano-4-dimethylamino-pyridinium tetrafluoroborate
PubChem CID 9881151
IUPAC Name 4-(dimethylamino)pyridin-1-ium-1-carbonitrile;tetrafluoroborate
SMILES F[B-](F)(F)F.CN(C)C1=CC=[N+](C=C1)C#N
Specifications
Melting Point 196°C to 200°C
Quantity 500 mg
Beilstein 5685616
Solubility Information Soluble in water,and acetonitrile.
Formula Weight 234.99
Percent Purity 98%
Chemical Name or Material 1-Cyano-4-(dimethylamino)pyridinium tetrafluoroborate