missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorooctadecane, 94%

Product Code. p-7027851
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11451367

Brand: Thermo Scientific Alfa Aesar B24900.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3386-33-2 | C18H37Cl | 288.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3386-33-2
Molecular Formula C18H37Cl
Molecular Weight (g/mol) 288.94
MDL Number MFCD00000960
InChI Key VUQPJRPDRDVQMN-UHFFFAOYSA-N
Synonym octadecyl chloride, octadecane, 1-chloro, n-octadecyl chloride, 1-chloroctadecane, chlorooctadecane, octadecane, chloro, unii-73h8vd533i, 1-chloranyloctadecane, stearylchloride, acmc-1cnf6
PubChem CID 18815
IUPAC Name 1-chlorooctadecane
SMILES CCCCCCCCCCCCCCCCCCCl

Specifications

Melting Point 20°C to 23°C
Density 0.849
Boiling Point 157°C to 158°C (1.5 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3(CH2)17Cl
Refractive Index 1.451
Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 288.95
Percent Purity 94%
Chemical Name or Material 1-Chlorooctadecane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.