Learn More
1-Chlorobutane, Sigma-Aldrich™
Product Code. 15828620
Change view
Click to view available options
Quantity:
100 mL
2.5 L
Unit Size:
100mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15828620 2.5 L 2.5 L
15808620 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15828620 Supplier Sigma Aldrich Supplier No. 8016402500
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 109-69-3
Grade Research
Molecular Formula CH3(CH2)3Cl
Molecular Weight (g/mol) 92.57
Specifications
Melting Point -123°C
Density 0.886 g/cm3 at 20°C
Boiling Point 79°C/1013 hPa
MDL Number MFCD00001009
Quantity 2.5 L
Synonym 1-Chlorobutane, n-Butyl chloride, 1-Chlorobutane