Learn More
Integra Biosciences™ 1-Channel Adapter
Product Code. 11777312
Change view
Click to view available options
Type:
Adapter for Disposable Tips
Adapter with Ejector for GripTips
Adapter, 150 mm
Adapter, 40 mm
Unit Size:
Each
Pack of 5
Product Code. Type unitSize
11777312 Adapter with Ejector for GripTips Each
10090272 Adapter, 150 mm Each
10319341 Adapter, 40 mm Each
12964417 Adapter for Disposable Tips Pack of 5
4 options
Product Code. 11777312
Brand: Integra Biosciences™ 159027
Only null left
Add to basket
Add to basket
For use with VACUBOY Hand Operator.
One-channel adaptor for use with VACUBOY hand operator. 
The VACUBOY hand operator can be used with VACUSAFE comfort vacuum system. Enables liquid residues to be disposed of reliably, precisely and conveniently:
  • With a precisely dosed volume
  • Without loss of drops
  • Without risk of contamination
  • Without causing tiredness
  • Energy saving, the manual operator - the heart of the system - can be operated as a single or 8 channel aspirator
  • All parts in contact with fluids are autoclavable
Specifications
Description 1-channel
Type Adapter with Ejector for GripTips
For Use With (Equipment) VACUBOY Hand Operator