Learn More
Thermo Scientific™ Cartridges for Multidrop™ Pico Digital Dispensers
Shop All Thermo Scientific ProductsDescription
The cartridges are designed for use with Multidrop™ Pico Digital Dispensers, dispensing volumes from 8 pL to 10 μL. Use the 1-channel cartridges with the Multidrop™ Pico 1, and the 4- or 8-channel standard or surfactant free cartridges with the Multidrop™ Pico 8 Surfactant Free (Pico 8 sf).
Features:
- Single-Channel Cartridges: Use the 1-channel cartridges with the Pico 1 to dispense DMSO, aqueous fluids with added surfactant, or master mixes. Dispensing volume: 11 pL – 10 μL..
- Standard Cartridges: Use the 4- or 8-channel standard cartridges with the Pico 8 sf to dispense DMSO, aqueous fluids with added surfactant, or master mixes. Dispensing volume: 11 pL – 10 μL.
- Surfactant Free Cartridges: Use the 4- or 8-channel surfactant free cartridges with the Pico 8 sf to dispense only aqueous fluids without surfactant. Dispensing volume: 8 pL – 10 μL.
- Contamination-Free Dispensing: Non-contact dispensing and single-use cartridges prevent carry-over and contamination.
Tekniske data
Tekniske data
| Product Type | Digital Dispenser Cartridge |
| Channels | 8 |
| Volume (Metric) | 8 pL to 10 μL |
| For Use With (Equipment) | Multidrop Pico 8 Surfactant Free Digital Dispenser |
| Model | Surfactant Free Cartridges, 8-channel 20 μL |
| No. per Pack | Case of 20 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?