Learn More
1-Butylpyrrolidine, TRC
Product Code. 30134649
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134649 100 mg 100mg
30137378 500 mg 500mg
30007111 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134649 Supplier Toronto Research Chemicals Supplier No. TRCB700948100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 767-10-2
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
Synonym N-Butylpyrrolidine, N-Butyltetrahydropyrrole
IUPAC Name 1-butylpyrrolidine
SMILES CCCCN1CCCC1
Tekniske data
Quantity 100 mg
InChI Formula InChI=1S/C8H17N/c1-2-3-6-9-7-4-5-8-9/h2-8H2,1H3
Formula Weight 127.1361
Chemical Name or Material 1-Butylpyrrolidine
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.