Learn More
1-Butanethiol, Sigma-Aldrich™
Product Code. 15814041
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15814041 500 mL 500mL
15804041 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15814041 Supplier Sigma-Aldrich Supplier No. 8015870500
 Restricted Item
Estimated Shipment: 23-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 109-79-5
Grade Research
Molecular Formula CH3(CH2)3SH
Molecular Weight (g/mol) 90.19
Specifications
Melting Point -116°C
Density 0.84 g/cm3 at 20°C
Boiling Point 97°C to 99°C/1013 hPa
MDL Number MFCD00004905
Quantity 500 mL
Synonym 1-Butanethiol, Butyl mercaptan, 1-Butanethiol