Learn More
1-(Bromomethyl)-3-(trifluoromethoxy)benzene, 97%, Thermo Scientific™
Product Code. 10344442
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10344442 1 g 1g
10280165 5 g 5g
10863761 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10344442 Supplier Thermo Scientific Maybridge Supplier No. SB01883DA
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 159689-88-0
Molecular Formula C8H6BrF3O
Molecular Weight (g/mol) 255.03
MDL Number MFCD00061271
InChI Key QSIVWRRHVXSDNE-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzyl bromide, 1-bromomethyl-3-trifluoromethoxy benzene, 3-trifluoromethoxybenzyl bromide, alpha-bromo-3-trifluoromethoxy toluene, 3-trifluoromethoxy benzylbromide, m-trifluoromethoxy benzyl bromide, 3-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-3-trifluoromethoxy-benzene, benzene, 1-bromomethyl-3-trifluoromethoxy
PubChem CID 737176
IUPAC Name 1-(bromomethyl)-3-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=CC(CBr)=C1
Specifications
CAS Min % 97.0
Density 1.5720g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CH2BrC6H4OCF3
Quantity 1 g
Formula Weight 255.03
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)-3-(trifluoromethoxy)benzene