Learn More
1-Bromo-4-nitrobenzene, 98%
Product Code. p-7023799
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
Product Code. Quantity unitSize
11434190 5 g 5g
11423288 25 g 25g
11433288 100 g 100g
11443288 500 g 500g
4 options
Product Code. 11434190
Brand: Thermo Scientific Alfa Aesar A17705.06
Request Bulk or Custom Format
CAS: 586-78-7 | C6H4BrNO2 | 202.01 g/mol
It can be employed as a syntheses material intermediate. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be employed as a syntheses material intermediate. It is also used as a pharmaceutical intermediate.
Solubility
Insoluble in water. Almost transparent in hot ethyl alcohol. Soluble in ether
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 586-78-7
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD00007280
InChI Key ZDFBKZUDCQQKAC-UHFFFAOYSA-N
Synonym 4-bromonitrobenzene, 4-nitrobromobenzene, p-bromonitrobenzene, benzene, 1-bromo-4-nitro, p-nitrobromobenzene, 4-nitrophenyl bromide, 4-bromo-1-nitrobenzene, p-nitrophenyl bromide, 1-bromo-4-nitro-benzene, ccris 3115
PubChem CID 11466
IUPAC Name 1-bromo-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Br)C=C1
Specifications
Melting Point 124°C to 127°C
Density 1.948
Boiling Point 254°C to 256°C
Quantity 5 g
UN Number UN3459
Beilstein 636964
Solubility Information Insoluble in water. Almost transparent in hot ethyl alcohol. Soluble in ether
Formula Weight 202.01
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-nitrobenzene