Learn More
1-Bromo-4-isopropylbenzene, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-4-isopropylbenzene |
| CAS | 586-61-8 |
| Molecular Formula | C9H11Br |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C9H11Br/c1-7(2)8-3-5-9(10)6-4-8/h3-7H,1-2H3 |
| SMILES | CC(C)c1ccc(Br)cc1 |
| IUPAC Name | 1-bromo-4-propan-2-ylbenzene |
| Molecular Weight (g/mol) | 199.09 |
| Formula Weight | 198.0044 |