Learn More
1-Bromo-4-fluoronaphthalene, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 341-41-3 |
| Molecular Formula | C10H6BrF |
| MDL Number | MFCD00051473 |
| Quantity | 500 g |
| SMILES | FC1=C2C=CC=CC2=C(Br)C=C1 |
| Molecular Weight (g/mol) | 225.06 |
| Percent Purity | 95% |
| Chemical Name or Material | 1-Bromo-4-fluoronaphthalene |