Learn More
1-Bromo-4-difluoromethoxy-2-fluorobenzene, 96%, Thermo Scientific™
Product Code. 15455356
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455356 250 mg 250mg
15426388 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15455356 Supplier Thermo Scientific Alfa Aesar Supplier No. H59931.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1242258-38-3
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.01
MDL Number MFCD18390897
InChI Key QWUYTYZPBMAYKP-UHFFFAOYSA-N
Synonym 1-bromo-4-difluoromethoxy-2-fluorobenzene, benzene,1-bromo-4-difluoromethoxy-2-fluoro, 1-bromo-4-difluoromethoxy-2-fluoro-benzene
PubChem CID 65203213
IUPAC Name 1-bromo-4-(difluoromethoxy)-2-fluorobenzene
SMILES FC(F)OC1=CC(F)=C(Br)C=C1
Specifications
Quantity 250 mg
Formula Weight 241.01
Percent Purity 96%
Chemical Name or Material 1-Bromo-4-difluoromethoxy-2-fluorobenzene
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.