Learn More
1-Bromo-2-naphthaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-2-naphthaldehyde |
| CAS | 3378-82-3 |
| Molecular Formula | C11H7BrO |
| Quantity | 2.5 g |
| Synonym | 1-Bromo-2-naphthalenecarboxaldehyde |
| InChI Formula | InChI=1S/C11H7BrO/c12-11-9(7-13)6-5-8-3-1-2-4-10(8)11/h1-7H |
| SMILES | O=CC1=CC=C2C=CC=CC2=C1Br |
| IUPAC Name | 1-bromo-2-naphthaldehyde |
| Molecular Weight (g/mol) | 235.08 |
| Formula Weight | 233.97 |