missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-3-methylaminopyrrolidine, 97%, Thermo Scientific™

Catalog Number 15515936
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
454712-26-6
C10H20N2O2
200.28
OKUCEQDKBKYEJY-UHFFFAOYSA-N
1-boc-3-methylaminopyrrolidine, tert-butyl 3-methylamino pyrrolidine-1-carboxylate, 1-boc-3-methylamino-pyrrolidine, 1-boc-3-methylamino pyrrolidine, 3-methylamino-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-pyrrolidinecarboxylic acid, 3-methylamino-, 1,1-dimethylethyl ester, s-1-boc-3-methylamino pyrrolidine, pubchem11215, acmc-1aqtj, acmc-20a85z
45089533
tert-butyl 3-(methylamino)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)NC
This item is not returnable. View return policy
454712-26-6
C10H20N2O2
200.28
OKUCEQDKBKYEJY-UHFFFAOYSA-N
1-boc-3-methylaminopyrrolidine, tert-butyl 3-methylamino pyrrolidine-1-carboxylate, 1-boc-3-methylamino-pyrrolidine, 1-boc-3-methylamino pyrrolidine, 3-methylamino-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-pyrrolidinecarboxylic acid, 3-methylamino-, 1,1-dimethylethyl ester, s-1-boc-3-methylamino pyrrolidine, pubchem11215, acmc-1aqtj, acmc-20a85z
45089533
tert-butyl 3-(methylamino)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)NC
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material 1-BOC-3-methylaminopyrrolidine
compliance-icons
Product Identifier
  • 1-BOC-3-methylaminopyrrolidine, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'