missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Boc-3-hydroxypyrrolidine, 97%, Thermo Scientific Chemicals

Catalog Number p-7030439
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
103057-44-9
C9H17NO3
187.239
MFCD04038535
APCBTRDHCDOPNY-UHFFFAOYSA-N
1-boc-3-hydroxypyrrolidine, 1-boc-3-pyrrolidinol, n-boc-3-hydroxypyrrolidine, n-boc-3-pyrrolidinol, 1-n-boc-3-hydroxypyrrolidine, 1-pyrrolidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-pyrrolidinol, 3-hydroxy-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-hydroxypyrrolidinecarboxylate, 1-tert-butoxycarbonyl-3-hydroxypyrrolidine
4416939
tert-butyl 3-hydroxypyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)O
This item is not returnable. View return policy
103057-44-9
C9H17NO3
187.239
MFCD04038535
APCBTRDHCDOPNY-UHFFFAOYSA-N
1-boc-3-hydroxypyrrolidine, 1-boc-3-pyrrolidinol, n-boc-3-hydroxypyrrolidine, n-boc-3-pyrrolidinol, 1-n-boc-3-hydroxypyrrolidine, 1-pyrrolidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-pyrrolidinol, 3-hydroxy-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-hydroxypyrrolidinecarboxylate, 1-tert-butoxycarbonyl-3-hydroxypyrrolidine
4416939
tert-butyl 3-hydroxypyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 60°C to 62°C
Quantity 5 g
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material (+/-)-1-Boc-3-hydroxypyrrolidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'