missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydryl-3-azetidinol hydrochloride, 95%, Thermo Scientific™

Product Code. 11465530
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465530 5 g 5g
11455530 1 g 1g
11475530 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465530 Supplier Thermo Scientific Alfa Aesar Supplier No. H25819.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90604-02-7
Molecular Formula C16H18ClNO
Molecular Weight (g/mol) 275.78
MDL Number MFCD00209447
InChI Key LCHTWRWPHBRTNO-UHFFFAOYSA-N
Synonym 1-benzhydrylazetidin-3-ol hydrochloride, 1-diphenylmethyl-3-hydroxyazetidine hydrochloride, 1-diphenylmethyl azetidin-3-ol hydrochloride, 1-benzhydryl-3-azetidinol hydrochloride, 1-benzhydrylazetan-3-ol-hcl, 1-benzhydrylazetidin-3-ol hcl, 1-benzhydryl azetidin-3-ol hcl, 1-diphenylmethyl-3-azetidinol hydrochloride, 1-benzhydrylazetan-3-ol hydrochloride, n-benzhydrylazetidin-3-ol hydrochloride
PubChem CID 2801726
IUPAC Name 1-benzhydrylazetidin-3-ol;hydrochloride
SMILES Cl.OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 172°C to 174°C
Quantity 5 g
Formula Weight 275.78
Percent Purity 95%
Chemical Name or Material 1-Benzhydryl-3-azetidinol hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.