Learn More
1-Aminoguanidinium hydrogen carbonate, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | 1-Aminoguanidinium hydrogen carbonate |
| CAS | 2582-30-1 |
| Melting Point | 125°C (decomposes) |
| Density | 1.56 g/cm³ at 20°C |
| Quantity | 25 kg |
| Synonym | 1-Aminoguanidinium hydrogen carbonate, Aminoguanidine bicarbonate, 1-Aminoguanidinium hydrogen carbonate |
| Molecular Weight (g/mol) | 136.11 |
| Grade | Not Available |