Learn More
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal is used in modification of polymer properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal is used in modification of polymer properties.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal is used in modification of polymer properties.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 301°C to 304°C |
Quantity | 1 g |
Beilstein | 1373325 |
Solubility Information | Insoluble in water. |
Formula Weight | 201.22 |
Percent Purity | 98% |
Chemical Name or Material | 1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal |