Learn More
(±)-1-Amino-2-propanol-1,1,2,3,3,3-d6, TRC
Product Code. 30322258
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322258 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30322258 Supplier Toronto Research Chemicals Supplier No. TRCA62813610MG
 Restricted Item
Estimated Shipment: 01-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (±)-1-Amino-2-propanol-1,1,2,3,3,3-d6
CAS 1219795-13-7
Molecular Formula C3 D6 H3 N O
Quantity 10 mg
Synonym 2-Propan-1,1,1,2,3,3-d6-ol, 3-amino- (ACI), 3-Amino-2-propan-1,1,1,2,3,3-d6-ol (ACI), (±)-1-Amino-2-propanol-1,1,2,3,3,3-d6, 1-Amino-2-propanol-1,1,2,3,3,3-d6, 1-Amino-2-propanol-D6
InChI Formula InChI=1S/C3H9NO/c1-3(5)2-4/h3,5H,2,4H2,1H3/i1D3,2D2,3D
SMILES [2H]C([2H])([2H])C([2H])(O)C([2H])([2H])N
IUPAC Name 1-amino-1,1,2,3,3,3-hexadeuteriopropan-2-ol
Molecular Weight (g/mol) 81.1466
Formula Weight 81.1061