missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanemethanol, 98%, Thermo Scientific Chemicals

Catalog Number 11460298
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
770-71-8
C11H18O
166.264
MFCD00074751
MDVGOOIANLZFCP-UHFFFAOYSA-N
1-adamantanemethanol, adamantan-1-ylmethanol, 1-adamantane methanol, 1-hydroxymethyl adamantane, adamantane-1-methanol, adamantan-1-yl-methanol, 1-hydroxymethyladamantane, tricyclo 3.3.1.1'3,7 dec-1-ylmethanol, adamantan-1-yl methanol, tricyclo 3.3.1.13,7 decanemethanol
64556
1-adamantylmethanol
C1C2CC3CC1CC(C2)(C3)CO
This item is not returnable. View return policy
770-71-8
C11H18O
166.264
MFCD00074751
MDVGOOIANLZFCP-UHFFFAOYSA-N
1-adamantanemethanol, adamantan-1-ylmethanol, 1-adamantane methanol, 1-hydroxymethyl adamantane, adamantane-1-methanol, adamantan-1-yl-methanol, 1-hydroxymethyladamantane, tricyclo 3.3.1.1'3,7 dec-1-ylmethanol, adamantan-1-yl methanol, tricyclo 3.3.1.13,7 decanemethanol
64556
1-adamantylmethanol
C1C2CC3CC1CC(C2)(C3)CO

1-Adamantanemethanol was used in the synthesis of axially disubstituted silicon(IV) phthalocyanines. It is used as a reagent in the synthesis of adamantane and noradamantane based histone deacetylase (HDAC) inhibitors for the treatment of cancer. It is also used as a reagent in the synthesis of (1-adamantyl)methyl glycidyl ether, a versatile building block for living polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanemethanol was used in the synthesis of axially disubstituted silicon(IV) phthalocyanines. It is used as a reagent in the synthesis of adamantane and noradamantane based histone deacetylase (HDAC) inhibitors for the treatment of cancer. It is also used as a reagent in the synthesis of (1-adamantyl)methyl glycidyl ether, a versatile building block for living polymerization.

Solubility
Insoluble in water. Solubility in methanol (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 115°C to 118°C
Quantity 1 g
Beilstein 1853339
Solubility Information Insoluble in water. Solubility in methanol (almost transparency).
Formula Weight 166.26
Percent Purity 98%
Chemical Name or Material 1-Adamantanemethanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'