Learn More
1,8-Dihydroxy-4,5-dinitroanthraquinone, 97%, Thermo Scientific™
Product Code. 11455997
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455997 250 g 250g
11435997 10 g 10g
11445997 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11455997 Supplier Thermo Scientific Alfa Aesar Supplier No. B25032.30
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 81-55-0
Molecular Formula C14H6N2O8
Molecular Weight (g/mol) 330.208
MDL Number MFCD00019149
InChI Key GJCHQJDEYFYWER-UHFFFAOYSA-N
Synonym 1,8-dihydroxy-4,5-dinitroanthraquinone, 4,5-dinitrochrysazin, 1,8-dinitro-4,5-dihydroxyanthraquinone, 9,10-anthracenedione, 1,8-dihydroxy-4,5-dinitro, unii-heo6302d6d, ccris 4796, 4,5-dihydroxy-1,8-dinitroanthraquinone, anthraquinone, 1,8-dihydroxy-4,5-dinitro, 1,8-dihydroxy-4,5-dinitro-9,10-anthracenedione, 1,8-dihydroxy-4,5-dinitro-anthraquinone
PubChem CID 3378440
IUPAC Name 1,8-dihydroxy-4,5-dinitroanthracene-9,10-dione
SMILES C1=CC(=C2C(=C1[N+](=O)[O-])C(=O)C3=C(C=CC(=C3C2=O)O)[N+](=O)[O-])O
Specifications
Melting Point 225°C (decomposition)
Quantity 250 g
Formula Weight 330.21
Percent Purity 97%
Chemical Name or Material 1,8-Dihydroxy-4,5-dinitroanthraquinone