Learn More
1,8-Dihydroxy-4,5-dinitro-9,10-anthracenedione, TRC
Specifications
Specifications
| Chemical Name or Material | 1,8-Dihydroxy-4,5-dinitro-9,10-anthracenedione |
| CAS | 81-55-0 |
| Molecular Formula | C14H6N2O8 |
| Quantity | 10 g |
| Synonym | 1,8-Dihydroxy-4,5-dinitro-anthraquinone, 4,5-Dinitro-chrysazin, 1,8-Dihydroxy-4,5-dinitro-9,10-anthraquinone, 1,8-Dihydroxy-4,5-dinitroanthracene-9,10-dione, 1,8-Dihydroxy-4,5-dinitroanthraquinone, 1,8-Dinitro-4,5-dihydroxyanthraquinone, 4,5-Dihydroxy-1,8-dinitroanthraquinone, 4,5-Dinitrochrysazin, ARDP 0006, NSC 81256 |
| InChI Formula | InChI=1S/C14H6N2O8/c17-7-3-1-5(15(21)22)9-11(7)14(20)12-8(18)4-2-6(16(23)24)10(12)13(9)19/h1-4,17-18H |
| SMILES | Oc1ccc(c2C(=O)c3c(C(=O)c12)c(O)ccc3[N+](=O)[O-])[N+](=O)[O-] |
| IUPAC Name | 1,8-dihydroxy-4,5-dinitroanthracene-9,10-dione |
| Molecular Weight (g/mol) | 330.21 |
| Formula Weight | 330.0124 |