Learn More
1,6-Naphthyridin-2(1H)-one, 97%, Thermo Scientific Chemicals
Catalog Number 11474570
Click to view available options
Quantity:
1 g
5 g
23616-29-7
C8H6N2O
146.15
MFCD07781179
ZQKMVHXJWJNEQG-UHFFFAOYSA-N
1,6-naphthyridin-2 1h-one, acmc-1cgmu, pyrido 3,2-c pyridone, hydroxy-1,6-naphthyridine, 1,6-naphthyridine-2-one, 2-hydroxy-1,6-naphthyridine, 1,6 naphthyridin-2-ol #, 1h-1,6 naphthyridin-2-one
589674
1H-1,6-naphthyridin-2-one
O=C1NC2=CC=NC=C2C=C1
23616-29-7
C8H6N2O
146.15
MFCD07781179
ZQKMVHXJWJNEQG-UHFFFAOYSA-N
1,6-naphthyridin-2 1h-one, acmc-1cgmu, pyrido 3,2-c pyridone, hydroxy-1,6-naphthyridine, 1,6-naphthyridine-2-one, 2-hydroxy-1,6-naphthyridine, 1,6 naphthyridin-2-ol #, 1h-1,6 naphthyridin-2-one
589674
1H-1,6-naphthyridin-2-one
O=C1NC2=CC=NC=C2C=C1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material 1,6-Naphthyridin-2(1H)-one