Learn More
1,6-Dicyanohexane, 99%, Thermo Scientific™
Product Code. 11490378
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490378 100 g 100g
11480378 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11490378 Supplier Thermo Scientific Alfa Aesar Supplier No. B24543.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 629-40-3
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00001981
InChI Key BTNXBLUGMAMSSH-UHFFFAOYSA-N
Synonym 1,6-dicyanohexane, suberonitrile, suberic acid dinitrile, suberodinitrile, unii-jnt1lxa02u, korksaeuredinitril german, jnt1lxa02u, hexamethylene di-isocyanide, hexane-1,6-dicarbonitrile, hexamethylenediisocyanic acid
PubChem CID 12385
IUPAC Name octanedinitrile
SMILES C(CCCC#N)CCC#N
Specifications
Melting Point -3°C
Density 0.93
Boiling Point 174°C to 176°C
Flash Point 175°C (347°F)
Refractive Index 1.4436
Quantity 100 g
UN Number UN3276
Beilstein 1744365
Formula Weight 136.2
Percent Purity 99%
Chemical Name or Material 1,6-Dicyanohexane