missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Isoquinolinesulfonyl)-2-methylpiperazine Dihydrochloride, TRC

Product Code. 30390049
Change view
Click to view available options
Quantity:
5 mg
250 mg
Unit Size:
5mg
Product Code. Quantity unitSize
30390049 250 mg -
30402602 5 mg 5mg
2 options
This item is not returnable. View return policy

Product Code. 30390049

Brand: Toronto Research Chemicals TRCI885000250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108930-17-2
Formula Weight 363.0575
InChI Formula InChI=1S/C14H17N3O2S.2ClH/c1-11-9-16-7-8-17(11)20(18,19)14-4-2-3-12-10-15-6-5-13(12)14;;/h2-6,10-11,16H,7-9H2,1H3;2*1H
IUPAC Name 5-(2-methylpiperazin-1-yl)sulfonylisoquinoline;dihydrochloride
Molecular Formula C14 H17 N3 O2 S . 2 Cl H
Molecular Weight (g/mol) 364.29
SMILES Cl.Cl.CC1CNCCN1S(=O)(=O)c2cccc3cnccc23
Chemical Name or Material 1-(5-Isoquinolinesulfonyl)-2-methylpiperazine dihydrochloride

Specifications

Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.