Learn More
1-[5-(Hydroxymethyl)-2-furyl]-2-methyl-1,2,3,4-tetrahydroisochinolin-4,8-diol, TRC
Product Code. 30011433
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011433 10 mg 10mg
30137875 5 mg 5mg
30003299 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011433 Supplier Toronto Research Chemicals Supplier No. TRCH82574510MG
 Restricted Item
Estimated Shipment: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1312706-18-5
Molecular Formula C15H17NO4
Molecular Weight (g/mol) 275.3
Synonym 1,2,3,4-Tetrahydro-1-[5-(hydroxymethyl)-2-furanyl]-2-methyl-4,8-isoquinolinediol;
SMILES OC1CN(C)C(C2=CC=C(CO)O2)C3=C1C=CC=C3O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C15H17NO4/c1-16-7-12(19)10-3-2-4-11(18)14(10)15(16)13-6-5-9(8-17)20-13/h2-6,12,15,17-19H,7-8H2,1H3
Formula Weight 275.1158
Chemical Name or Material 1-[5-(Hydroxymethyl)-2-furyl]-2-methyl-1,2,3,4-tetrahydroisochinolin-4,8-diol