missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diazabicyclo[4.3.0]non-5-ene, Sigma-Aldrich™

Product Code. 15849060
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15849060 100 mL 100mL
15839060 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15849060 Supplier Sigma Aldrich Supplier No. 8036030100
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3001-72-7
Grade Research
Molecular Weight (g/mol) 124.18

Specifications

Density 1.04 g/cm3 at 20°C
Boiling Point 110°C/20 hPa
MDL Number MFCD00005554
Quantity 100 mL
Synonym 1,5-Diazabicyclo[4.3.0]non-5-ene, Pyrrolidino[1,2:a]1,4,5,6-tetrahydropyrimidine, DBN, 1,5-Diazabicyclo[4.3.0]non-5-ene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.