Learn More
1-(5-Bromo-2-hydroxyphenyl)ethanone, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(5-Bromo-2-hydroxyphenyl)ethanone |
| CAS | 1450-75-5 |
| Molecular Formula | C8 H7 Br O2 |
| Quantity | 1 g |
| Synonym | Acetophenone, 5'-bromo-2'-hydroxy- (6CI,7CI,8CI), 1-(5-Bromo-2-hydroxyphenyl)ethan-1-one, 2-Acetyl-4-bromophenol, 2'-Hydroxy-5'-bromoacetophenone, 3-Bromo-6-hydroxyphenyl methyl ketone, 5-Bromo-2-hydroxyacetophenone, 5'-Bromo-2'-hydroxyacetophenone, 5'-Bromo-o-hydroxyacetophenone, NSC 46621 |
| InChI Formula | InChI=1S/C8H7BrO2/c1-5(10)7-4-6(9)2-3-8(7)11/h2-4,11H,1H3 |
| SMILES | CC(=O)c1cc(Br)ccc1O |
| IUPAC Name | 1-(5-bromo-2-hydroxyphenyl)ethanone |
| Molecular Weight (g/mol) | 215.044 |
| Formula Weight | 213.963 |