Learn More
Alfa Aesar™ 1-(4-Methoxyphenyl)-1H-1,2,4-triazole, 99%
Catalog Number 15434017
Click to view available options
Quantity:
5 g
25 g
68377-33-3
C9H9N3O
175.191
MFCD12407079
HXCCOAUMJBFQCC-UHFFFAOYSA-N
1-4-methoxyphenyl-1h-1,2,4-triazole, 1-4-methoxyphenyl-1,2,4-triazole, 1-4-methoxyphenyl-1h-1,2,4 triazole, 4-n-methoxyphenyl-1,2,4-triazole, 1-4-methoxy-phenyl-1h-1,2,4 triazole
11084390
1-(4-methoxyphenyl)-1,2,4-triazole
COC1=CC=C(C=C1)N2C=NC=N2
68377-33-3
C9H9N3O
175.191
MFCD12407079
HXCCOAUMJBFQCC-UHFFFAOYSA-N
1-4-methoxyphenyl-1h-1,2,4-triazole, 1-4-methoxyphenyl-1,2,4-triazole, 1-4-methoxyphenyl-1h-1,2,4 triazole, 4-n-methoxyphenyl-1,2,4-triazole, 1-4-methoxy-phenyl-1h-1,2,4 triazole
11084390
1-(4-methoxyphenyl)-1,2,4-triazole
COC1=CC=C(C=C1)N2C=NC=N2
Melting Point 96°C to 98°C
Quantity 5 g
Formula Weight 175.19
Percent Purity 99%
Chemical Name or Material 1-(4-Methoxyphenyl)-1H-1,2,4-triazole