Learn More
Motic™ 1/4 Lambda Plate
Product Code. 15415282
Change view
Click to view available options
For Use With (Equipment):
Polarizing microscope
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15415282 Polarizing microscope Each
1 options
Product Code. 15415282
Brand: Motic™ 1101006700162
Only null left
Add to basket
Add to basket
Assess optical path differences in birefringent specimens. Motic™ 1/4 Lambda Plate can be used to detect optical path differences of (plus or minus) one-half wavelength and produce circularly polarized light by inserting it into the optical path above the polarizer.
  • Commonly employed for qualitative retardation measurements of petrographic specimens (rock and mineral thin sections) or other birefringent materials whose retardation value falls within the wedge limits.
  • Turn plane-polarized light into circularly polarized light and vice-versa.
Specifications
Type Lambda Plate, 1/4
For Use With (Equipment) Polarizing microscope