Learn More
1-(4-Fluorophenyl)biguanide hydrochloride, 98%, Thermo Scientific™
Catalog Number 11361857
Click to view available options
Quantity:
1 g
5 g
16018-83-0
C8H11ClFN5
231.659
MFCD00051986
HBYJTLZNKNUTCP-UHFFFAOYSA-N
1-4-fluorophenyl biguanide hydrochloride, 1-carbamimidamido-n-4-fluorophenyl methanimidamide hydrochloride, biguanide, 1-p-fluorophenyl-, monohydrochloride, acmc-1bs1v, 1-diaminomethylidene-2-4-fluorophenyl guanidine hydrochloride, 1-4-fluorophenyl biguanide hydrochlorid, 1-4-fluorophenyl biguanidine hydrochloride, n-4-fluorophenyl imidodicarbonimidic diamide hydrochloride, 4-fluoroanilino imino methyl amino methanimidamide hydrochloride
11424753
1-(diaminomethylidene)-2-(4-fluorophenyl)guanidine;hydrochloride
C1=CC(=CC=C1N=C(N)N=C(N)N)F.Cl
16018-83-0
C8H11ClFN5
231.659
MFCD00051986
HBYJTLZNKNUTCP-UHFFFAOYSA-N
1-4-fluorophenyl biguanide hydrochloride, 1-carbamimidamido-n-4-fluorophenyl methanimidamide hydrochloride, biguanide, 1-p-fluorophenyl-, monohydrochloride, acmc-1bs1v, 1-diaminomethylidene-2-4-fluorophenyl guanidine hydrochloride, 1-4-fluorophenyl biguanide hydrochlorid, 1-4-fluorophenyl biguanidine hydrochloride, n-4-fluorophenyl imidodicarbonimidic diamide hydrochloride, 4-fluoroanilino imino methyl amino methanimidamide hydrochloride
11424753
1-(diaminomethylidene)-2-(4-fluorophenyl)guanidine;hydrochloride
C1=CC(=CC=C1N=C(N)N=C(N)N)F.Cl
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Melting Point 212°C to 216°C
Quantity 1 g
Formula Weight 231.66
Percent Purity 98%
Chemical Name or Material 1-(4-Fluorophenyl)biguanide hydrochloride