missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dioxane, Reagent Grade, ≥99%, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 mL
2.5 L
10 L
20 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
10 L
2.5 L
20 L
500mL
Pack of 6
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| pH | 6 to 8 (at 500.00g/L 20°C) |
| Melting Point | 12°C |
| Color | Colorless |
| Density | 1.034g/mL at 25°C |
| Boiling Point | 100°C to 102°C (1013 hPa) |
| Quantity | 2.5 L |
| Explosion Limit | 0.22 |
| Flash Point | 12°C (53.6°F) |
| Ignition Point | 356°F |
| Odor | Aromatic |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction