missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, Reagent Grade, ≥99%, Honeywell™

Product Code. 15657800 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
10 L
20 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
10 L
2.5 L
20 L
500mL
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15657800 10 L Stainless Steel Can 10 L
15687800 500 mL Glass Bottle 500mL
15667800 2.5 L Glass Bottle 2.5 L
15697800 6 x 1 L Glass Bottle Pack of 6
15677800 20 L Stainless Steel Can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15657800 Supplier Honeywell Research Chemicals Supplier No. D20186310L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

pH 6 to 8 (at 500.00g/L 20°C)
Melting Point 12°C
Color Colorless
Density 1.034g/mL at 25°C
Boiling Point 100°C to 102°C (1013 hPa)
Quantity 10 L
Explosion Limit 0.22
Flash Point 12°C (53.6°F)
Ignition Point 356°F
Odor Aromatic
Packaging Stainless Steel Can
Physical Form Liquid
Vapor Pressure 27mmHg (20°C), 40mmHg (25°C)
UN Number UN1165
Beilstein 102551
Solubility Information Fully miscible in water
Formula Weight 88.11g/mol
Percent Purity ≥99%
Chemical Name or Material 1,4-Dioxane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.