Learn More
1,4-Diiodobenzene-13C6, TRC
Specifications
Specifications
| Chemical Name or Material | 1,4-Diiodobenzene-13C6 |
| CAS | 1246817-17-3 |
| Molecular Formula | 13C6H4I2 |
| Quantity | 50 mg |
| Synonym | 1,4-Iodobenzene-13C6, 4-Iodophenyl Iodide-13C6, NSC 6297-13C6, p-Diiodobenzene13C6 |
| InChI Formula | InChI=1S/C6H4I2/c7-5-1-2-6(8)4-3-5/h1-4H/i1+1,2+1,3+1,4+1,5+1,6+1 |
| SMILES | I[13c]1[13cH][13cH][13c](I)[13cH][13cH]1 |
| Molecular Weight (g/mol) | 335.86 |
| Formula Weight | 335.8604 |