Learn More
1,4-Dihydroxyanthraquinone, Sigma-Aldrich™
Product Code. 15864511
Change view
Click to view available options
Quantity:
5 g
250 g
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15864511 250 g 250g
15854511 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15864511 Supplier Sigma Aldrich Supplier No. 8202590250
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 81-64-1
Grade Research
Molecular Weight (g/mol) 240.21
Specifications
Melting Point 195°C to 198°C
Boiling Point 450°C/1013 hPa
MDL Number MFCD00001209
Quantity 250 g
Synonym 1,4-Dihydroxyanthraquinone, Quinizarin, 1,4-Dihydroxyanthraquinone