Learn More
1,4-Dibromo-2,5-dimethoxybenzene, 98+%
Product Code. 11428697
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428697 50 g 50g
11418697 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11428697 Supplier Thermo Scientific Alfa Aesar Supplier No. A19858.18
Request Bulk or Custom Format
CAS: 2674-34-2 | C8H8Br2O2 | 295.958 g/mol
It is employed as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2674-34-2
Molecular Formula C8H8Br2O2
Molecular Weight (g/mol) 295.958
MDL Number MFCD00025694
InChI Key CHCLRVOURKGRSW-UHFFFAOYSA-N
Synonym 2,5-dibromo-1,4-dimethoxybenzene, 1,4-dibromo-2,5-dimethoxy-benzene, 2,5-dibromohydroquinone dimethyl ether, benzene, 1,4-dibromo-2,5-dimethoxy, pubchem16840, acmc-209gsv, maybridge1_005131, ksc496i0h, 2,5-dibromo-4-methoxyanisole, 1,4-dibromo-3,6-dimethoxybenzene
PubChem CID 231240
IUPAC Name 1,4-dibromo-2,5-dimethoxybenzene
SMILES COC1=CC(=C(C=C1Br)OC)Br
Specifications
Melting Point 144°C to 148°C
Quantity 50 g
Beilstein 1953103
Solubility Information Insoluble in water.
Formula Weight 295.96
Percent Purity ≥98%
Chemical Name or Material 1,4-Dibromo-2,5-dimethoxybenzene