missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diacetoxybutane, 98%, Thermo Scientific™

Product Code. 11458816
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458816 100 g 100g
11448816 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458816 Supplier Thermo Scientific Alfa Aesar Supplier No. A16978.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-67-1
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00014984
InChI Key XUKSWKGOQKREON-UHFFFAOYSA-N
Synonym 1,4-diacetoxybutane, 1,4-butanediol diacetate, tetramethylene diacetate, 1,4-butanediol, diacetate, butylene glycol diacetate, tetramethylene acetate, 1,4-butylene glycol diacetate, 1,4-butanediol, 1,4-diacetate, butane-1,4-diol diacetate, 1,4-butylene diacetate
PubChem CID 69410
ChEBI CHEBI:87345
IUPAC Name 4-acetyloxybutyl acetate
SMILES CC(=O)OCCCCOC(=O)C

Specifications

Melting Point 12°C to 15°C
Density 1.048
Boiling Point 230°C
Refractive Index 1.424
Quantity 100 g
Beilstein 1768757
Formula Weight 174.2
Percent Purity 98%
Chemical Name or Material 1,4-Diacetoxybutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.