Learn More
1-[(4-Chlorophenyl)methyl]-2,3-dihydroimidazo[1,2-a]pyridinium Chloride, Mikromol
Product Code. 30463431
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
100mg
25mg
Product Code. 30463431
Brand: Mikromol MM1894.0300254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-[(4-Chlorophenyl)methyl]-2,3-dihydroimidazo[1,2-a]pyridinium Chloride, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-[(4-Chlorophenyl)methyl]-2,3-dihydroimidazo[1,2-a]pyridinium Chloride
CAS 63111-28-4
Analyte or Component Names 1-[(4-Chlorophenyl)methyl]-2,3-dihydroimidazo[1,2-a]pyridinium Chloride
Shipping Condition Room Temperature
Molecular Formula C14 H14 Cl N2 . Cl
Certifications/Compliance ISO 17025
Synonym 1-[(4-Chlorophenyl)methyl]-2,3-dihydroimidazo[1,2-a]pyridinium Chloride, 1-[(4-Chlorophenyl)methyl]-2,3-dihydro-imidazo[1,2-a]pyridinium Chloride (1:1), 1-(4-Chlorobenzyl)-2,3-dihydro-1H-imidazol[1,2-a]pyridinium Chloride
InChI Formula InChI=1S/C14H14ClN2.ClH/c15-13-6-4-12(5-7-13)11-17-10-9-16-8-2-1-3-14(16)17;/h1-8H,9-11H2;1H/q+1;/p-1
Solution Type Neat
SMILES [Cl-].Clc1ccc(CN2CC[n+]3ccccc23)cc1
IUPAC Name 1-[(4-chlorophenyl)methyl]-2,3-dihydroimidazo[1,2-a]pyridin-4-ium;chloride
Molecular Weight (g/mol) 281.18
Quantity 4 x 25 mg
Formula Weight 280.0534 g/mol