Learn More
1-(4-Chlorophenyl)-2,2,2-trifluoroethanone, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 321-37-9 |
| Molecular Formula | C8H4ClF3O |
| MDL Number | MFCD00018686 |
| Quantity | 1 g |
| SMILES | FC(F)(F)C(=O)C1=CC=C(Cl)C=C1 |
| Molecular Weight (g/mol) | 208.57 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-(4-Chlorophenyl)-2,2,2-trifluoroethanone |