Learn More
1,4-Benzenediacrylic acid, 98%, Thermo Scientific Chemicals
Product Code. 15478789
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 15478789
Brand: Thermo Scientific Alfa Aesar L01141.14
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16323-43-6
Molecular Formula C12H10O4
Molecular Weight (g/mol) 218.208
MDL Number MFCD00002698
InChI Key AAFXQFIGKBLKMC-KQQUZDAGSA-N
Synonym 1,4-benzenediacrylic acid, 1,4-phenylenediacrylic acid, p-phenylenediacrylic acid, p-benzenediacrylic acid, unii-0f7e4o8q8y, 3,3'-1,4-phenylene bis-2-propenoic acid, 3,3'-p-phenylene diacrylic acid, 2e-3-4-1e-2-carboxyeth-1-en-1-yl phenyl prop-2-enoic acid, 2'e-3,3'-benzene-1,4-diylbisprop-2-enoic acid, 3,3'-benzene-1,4-diylbisprop-2-enoic acid
PubChem CID 759280
IUPAC Name (E)-3-[4-[(E)-2-carboxyethenyl]phenyl]prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)C=CC(=O)O
Specifications
Melting Point >300°C
Quantity 25 g
Beilstein 2579464
Formula Weight 218.21
Percent Purity 98%
Chemical Name or Material 1,4-Benzenediacrylic acid