Learn More
Trajan Scientific and Medical™ 1/32 in. O.D. PEEKsil™ Tubing
Product Code. 15278773
Change view
Click to view available options
Overall Length (Metric):
50 mm
100 mm
150 mm
250 mm
500 mm
Diameter (Metric) Inner:
0.025 mm
0.05 mm
0.075 mm
0.1 mm
0.15 mm
Unit Size:
Each
Pack of 2
Product Code. Overall Length (Metric) Diameter (Metric) Inner unitSize
15278773 250 mm 0.15 mm Pack of 2
15208753 250 mm 0.025 mm Pack of 2
15218753 500 mm 0.025 mm Pack of 2
15208763 250 mm 0.05 mm Pack of 2
15228773 100 mm 0.1 mm Pack of 2
15258773 500 mm 0.1 mm Pack of 2
15248773 250 mm 0.1 mm Pack of 2
12726248 500 mm 0.075 mm Each
15288773 500 mm 0.15 mm Pack of 2
12706248 150 mm 0.075 mm Each
12716248 250 mm 0.075 mm Pack of 2
15218773 50 mm 0.1 mm Pack of 2
15238773 150 mm 0.1 mm Pack of 2
15268773 100 mm 0.15 mm Pack of 2
15278743 50 mm 0.025 mm Pack of 2
15288743 100 mm 0.025 mm Pack of 2
15298743 150 mm 0.025 mm Pack of 2
17 options
Product Code. 15278773
Brand: Trajan Scientific and Medical™ 0624345
Only null left
Add to basket
Add to basket
A mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems
  • Fused silica internal wall provides smooth surface, free of imperfections
  • PEEKsil compatible with most organic solvents and resistant to strong acids
  • Effective pH range of 0 to 10
  • Flexibility ensures easy connection between columns, detectors and injectors
    Specifications
    Type Tubing
    Includes 2/Pk.
    Diameter (English) Outer 1/32 in.
    Diameter (Metric) Inner 0.15 mm
    Overall Length (Metric) 250 mm