Learn More
1,3-thiazol-2-ylmethylamine, 97%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 5 g |
| Formula Weight | 150.63 |
| Percent Purity | 97% |
| Chemical Name or Material | 1, 3-Thiazol-2-ylmethylamine hydrochloride |