Learn More
1,3-Propanediol, Sigma-Aldrich™
Product Code. 15819490
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15819490 1 L 1 L
15809490 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15819490 Supplier Sigma Aldrich Supplier No. 8074811000
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 504-63-2
Grade Research
Molecular Formula HO(CH2)3OH
Molecular Weight (g/mol) 76.09
Specifications
Melting Point -32°C
Density 1.05 g/cm3 at 20°C
Boiling Point 213°C to 215°C/1013 hPa
MDL Number MFCD00002949
Quantity 1 L
Synonym 1,3-Propanediol, Trimethylene glycol, 1,3-Propylene glycol, 1,3-Propanediol